Taq DNA Polymerase Blend, 500 U, Extender™
N638-KITEA 187 EUR
VWRCN638-KIT
Taq DNA Polymerase Blend, 500 U, Extender™
Extender™ Taq DNA Polymerase is a high fidelity, thermostable enzyme blend optimised to generate high product yields and purity. This polymerase blend includes 3' to 5’ proofreading activity for accurate template replication. Ideal for amplification of critical templates containing GC-rich regions, palindromes or multiple repeats.
  • Superior yields and purity
  • Excellent for hard-to-copy templates
  • Amplified products include a mixture of blunt ends and 3'dA overhangs
  • Wide tolerance for Mg2+, salt concentration, pH and template contaminants
Delivery information: Kit includes 1× Extender™ Taq DNA Polymerase Blend, 500 U (5 U/µl), 1× Extender™ Taq DNA Polymerase Buffer A, 10X (without MgCl2), 1× Extender™ Taq DNA Polymerase Buffer S, 10X (with MgCl2), and 1× Extender Polymerase MgCl2, 50 mM.