DNA amplification kits, PCR Kit with Taq Polymerase
N555-KITEA 244 EUR
VWRCN555-KIT
DNA amplification kits, PCR Kit with Taq Polymerase
PCR Kit with Taq Polymerase is a DNA amplification kit containing all reagents for PCR reactions, except template and primer.
  • Amplify DNA templates with high sensitivity
  • Amplify single-copy and low abundance templates
  • Nuclease-free, high purity reagents
The PCR Kit with Taq DNA Polymerase includes separate solutions of all material needed for a PCR reaction, including dNTPs, reaction buffer with and without MgCl2, 25 mM MgCl2, Taq Polymerase, and sterile water.
Delivery information: Kits include 10 mM of each dNTP, 50X, 10X, Reaction Buffer with MgCl2, 10X Reaction Buffer without MgCl2, 25 mM MgCl2, Taq DNA Polymerrase, 500 U (5 U/µl), and nuclease-free sterile water (2×1,5 ml) .