Hand cleanser dispenser, GrittyFOAM®
Supplier: SC Johnson
Deb Stoko® GrittyFOAM®
115-2091EA 48.6 EUR
115-2091
Hand cleanser dispenser, GrittyFOAM®
Soap Dispensers and Lotion Soap Dispensers
This dispenser for exclusive use with GrittyFOAM® cartridges, is ideal for use in light and heavy industrial environments with high usage, such as construction sites and repair shops.
  • Patented foam technology converts hand cleaning liquid into creamy foam
  • Silver-based antimicrobial agent BioCote® inhibits the growth of bacteria and moulds on the surface of the dispenser
  • Push-button enables one handed operation
  • Made of durable plastic
  • Wall-mounted
For use with Solopol® GrittyFOAM® hand cleanser.