Centrifuge, benchtop, refrigerated, Sorvall™ ST 16R
Supplier: Thermo Scientific
HERA75004380EA 8420 EUR
HERA75004380
Centrifuge, benchtop, refrigerated, Sorvall™ ST 16R
The Thermo Scientific™ Sorvall™ ST 16 centrifuge is compatible with a wide range of rotors.
  • Motorised lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
  • Auto-Lock™ III Rotor System: Secure locking system allows easy push-button installation and exchange of rotors; saves time switching between applications
  • Easy access to rotor chamber for quick cleaning
  • ClickSeal™ Bucket Sealing System: Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • SMARTSpin™ Technology: Advanced rotor management system maximises acceleration, braking and residual load imbalance for each rotor and bucket option
  • CFC-free refrigeration system, pre-cooling function with direct button
Ideal for cell culture and blood processing applications.
Ordering information: Supplied without rotors and adapters which need to be ordered separately.