Centrifuge, floor-standing, Sorvall™ Evolution™ RC superspeed
Supplier: Thermo Fisher Scientific
Sorvall™
SORV728411EA 44520 EUR
SORV728411
Centrifuge, floor-standing, Sorvall™ Evolution™ RC superspeed
Centrifuges Floor-Standing Centrifuges
This centrifuge delivers simplified sample handling, reduced overall spin times and superior ergonomics.
  • User-friendly instrument panel and intuitive software design
  • RunBrowser provides quick set-up of frequently-used protocols
  • Achieves high performance levels automatically, minimizing the amount of user attention required
  • DuraFlex gyro mechanism accommodates unbalanced rotors to prevent damage to the drive shaft or motor
  • Compact, ergonomic design: Deck is only 850 mm from the ground, making the rotor chamber and control panel easily accessible
Ordering information: A broad range of rotors and accessories is available separately.