PCR Taq mixture
Supplier: Omega Bio-Tek
TQ2200-00EA 29.9 EUR
OMEGTQ2200-00 OMEGTQ2200-01
PCR Taq mixture
This 2X Taq Master Mix is a premixed 2X concentrated solution of Taq DNA polymerase, reaction buffer, MgCl2, dNTPs, stabiliser, and enhancer.
  • Contains all components for PCR, except dNA template and primers
  • Mixture is optimised for consistent and efficient routine PCR amplifications
  • Can amplify up to 8 kb fragments from lambda DNA
For a 50 µl reaction, 25 µl of 2X Taq Master Mix is added to primers, DNA template, and PCR-quality water. In addition, 2X Taq Master Mix (with dye) can be used to directly load the PCR products onto an agarose gel without the need to add a gel loading buffer.