Taq DNA polymerase
Supplier: Omega Bio-Tek
TQ2100-02EA 590 EUR
TQ2100-02 TQ2100-01 TQ2100-00
Taq DNA polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq DNA polymerase is a thermostable enzyme that can withstand prolonged incubation at temperatures up to 95°C without significant loss of activity.
  • Consists of a single polypeptide with a molecular weight of 94 kDa
  • Has a 5´-3´ DNA polymerase activity and a 5´-3´ exonuclease activity
  • Concentration is 100 mM, and it is ready to use for PCR or other reactions