Chemiluminescent substrate for HRP, North2South™
Supplier: Thermo Scientific
Pierce
PIER17295EA 448 EUR
PIER17295
Chemiluminescent substrate for HRP, North2South™
Electrophoresis Reagents Blotting Reagents
North2South™ Chemiluminescent Substrate is used for detecting nucleic acids in northern and Southern blot applications.
This system uses an enhanced luminol substrate for horseradish peroxidase (HRP) that greatly reduces processing and film exposure time. Post-hybridisation processing has been reduced from the standard 2,5 hours to 1 hour. Film exposure times range from 30 seconds to 10 minutes with the substrate emitting light with relatively constant intensity for up to 6 hours, allowing for multiple exposures.
Delivery information: Kit includes luminol/enhancer solution (50 ml) and stable peroxide solution (50 ml).