Cosmic calf serum, New Zealand origin, HyClone™
Supplier: HyClone products (Cytiva)
HYCLSH30413.02HIEA 127 EUR
HYCLSH30413.02HI HYCLSH30413.03 HYCLSH30413.02IR HYCLSH30413.02
Cosmic calf serum, New Zealand origin, HyClone™
HyClone™ Cosmic calf serum is fortified with iron and growth promoting factors.
  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing