O-GlcNAc Western Blot Detection Kit

Supplier: Thermo Scientific
Pierce
PIER24565EA 0 EUR
PIER24565
O-GlcNAc Western Blot Detection Kit
Pierce™ O-GlcNAc Western Blot Detection Kit contains a specific antibody for the O-GlcNAc post-translational modification (PTM), as well as detection reagents needed to perform a complete detection experiment.

  • Highly specific monoclonal antibody against O-GlcNAc modification
  • Speed and sensitivity of chemiluminescent detection
  • Specific affinity for the beta-O-linked serine or threonine GlcNAc modification

Delivery information: The kit contains sufficient reagents for approximately 10 mini blots:
Part A:
M-PER™ Mammalian Protein Extraction Reagent, 25 ml
Dilution Buffer (also functions as Blocking Buffer) (10X), 2×50 ml
BupH™ Phosphate Buffered Saline, 17 packs
Surfact-Amps™ 20 (10% Tween™ 20 Detergent), 3×10 ml
Goat Anti-Mouse IgM (µ), HRP Conjugate, 75 µg
SuperSignal™ West Dura Luminol/Enhancer Solution, 50 ml
SuperSignal™ West Dura Stable Peroxide Solution, 50 ml
Part B:
Anti-O-GlcNAc Monoclonal Antibody (MAb CTD 110.6), 100 µl
O-GlcNAc Western Blot Positive Control (O-GlcNAc-modified BSA), 100 ng
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR