Durometers, shore hardness tester HB
Supplier: Kern & Sohn
KERNHBA100-0EA 96.9 EUR
KERNHBA100-0 KERNHBD100-0 KERNHB0100-0
Durometers, shore hardness tester HB
Durometers
Handheld durometer, HB, with drag indicator.
  • Typical application in the measurement of penetration
  • Shore A: rubber, elastomers, neoprene, silicone, vinyl, soft plastics, felt, leather and similar material
  • Shore D: plastics, formica, epoxides, plexiglass
  • Shore A0: foam, sponge
Max mode holds the maximum mode in display and point mode shows one instant value. This can be attached to the test stands SAUTER TI-A0 (for Shore A and A0) or TI-D (for Shore D).
Delivery information: Supplied in a wooden carrying case.