Glycerol-based agarose gel loading dye (6X)
Supplier: Thermo Fisher Scientific
J63429.ADEA 73.2 EUR
ALFAJ63429.AD
Glycerol-based agarose gel loading dye (6X)
Glycerol-based bromophenol-free agarose gel loading dyes.
Ordering information: Contains xylene cyanol FF (0,25 %) and glycerol (30 %).
Caution: Does not contain bromophenol blue.