Direct Methanol Fuel Cell (DMFC) cathodes
45368.KSEA 201 EUR
ALFA45368.KS
Direct Methanol Fuel Cell (DMFC) cathodes
Direct-methanol fuel cells (DMFC) cathodes find application in direct methanol fuel cell, which is used for powering large vehicles, smaller vehicles such as forklifts and tuggers and consumer goods such as mobile phones, digital cameras, laptops. Further, they can be used in military as power for man-portable tactical equipment, battery chargers and autonomous power for test and training instrumentation.
Incompatible with oxidising agents and organic materials.
Appearance: Black
Odour: Odourless