Ashing furnaces with fold down or lift door, LV and LVT series
Supplier: Nabertherm
455-1944EA 3530 EUR
455-1944 455-1950 455-1974 455-1986 455-1998 455-1938 455-2010 455-2016 455-1968 455-1962 455-1992 455-2008
Ashing furnaces with fold down or lift door, LV and LVT series
Ovens Furnaces (Ashing and Muffle Ovens)
These models are especially designed for ashing in the laboratory. Units feature rust-free, textured stainless steel exteriors. A special air intake and exhaust system allows air exchange of more than 6 times per minute. Incoming air is pre-heated to ensure a good temperature uniformity. Models have Tmax 1100 °C, and feature a silent solid state power control relay.
  • Heating on two sides by means of ceramic heating plates, module lining is highly durable, high performance cured vacuum fibre
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and is easy to replace
  • Double walled casing for low external temperatures and stability
  • Optional fold down door (LV models) which can be used as work platform or lift door (LVT models) with hot surface facing away from the operator
  • Air exchanged more than 6 times per minute
  • Non-classified insulation avoids the potential risk of cancer
Delivery information: Supplied with Standard B 510 controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions or optional C 550 controller with LC display, 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. All units are supplied with plugs and these models require a 230 V single phase power supply.
Caution: Maximum continuous operating temperature should be 50 °C below the max. temp. of the model shown in the table above.