Labels for NMR/EPR tubes, pressure sensitive, SP Wilmad-LabGlass
Supplier: SP Industries
WILMWGL-5EA 31.8 EUR
WILMWGL-5
Labels for NMR/EPR tubes, pressure sensitive, SP Wilmad-LabGlass
These labels are a handy alternative to marking NMR or EPR sample tubes. Each fits the circumference of the tube precisely with no overlapping to guarantee symmetry in spinning experiments.