Sequencing Kit, dedicated height
Supplier: CBS Scientific
730-0044EA 1530 EUR
CBSCDH-400-20
Sequencing Kit, dedicated height
Dedicated Height Sequencing Kit features a bar clamp, safety-interlocking lids, detachable buffer reservoirs for ease in cleaning and a built-in aluminum heat dispersion plate. Includes 4 mm×34-well sharktooth comb, 0,4×420 mm (h) spacer set, and 420 mm (h) glass plate set.