Flasks, pear shaped with standard ground joints, PYREX®
Supplier: Corning
Corning®
CORN4315-25
Flasks, pear shaped with standard ground joints, PYREX®
Flasks Pear Shaped Flasks
PYREX® borosilicate glass, short neck.
  • High thermal shock resistance
  • High chemical durability
  • Provide contamination-free results
The wall thicknesses of the flasks are carefully controlled to provide the proper balance between thermal expansion and mechanical strength. Their normal working temperature is 230 °C. Different adapters are used to connect flasks with different size joints, thus allowing numerous combinations for diverse assemblies.