Automatic test and calibration stations, MicroDock II
Supplier: BW Technologies
BWTEDOCK221G00GEA 1430 EUR
BWTEDOCK221G00G BWTEDOCK201N00G BWTEDOCK221C1P03G BWTEDOCK201P00G BWTEDOCK201J00G BWTEDOCK221C1N03G BWTEDOCK221E00G BWTEDOCK221D1P00G BWTEDOCK221D1N00G BWTEDOCK2200G BWTEDOCK201F00G BWTEDOCK231D1P00G BWTEDOCK201H00G BWTEDOCK231D00G BWTEDOCK231J00G BWTEDOCK221J1P00G BWTEDOCK231J1P00G BWTEDOCK221E03G BWTEDOCK251C1D1K00 BWTEDOCK201M00G BWTEDOCK221C1P00G BWTEDOCK231C1N00G BWTEDOCK201K00G BWTEDOCK231C1D1P00 BWTEDOCK221C1N00G BWTEDOCK231C1K03G BWTEDOCK201C1P00G BWTEDOCK221N00G BWTEDOCK221D00G BWTEDOCK221E01G BWTEDOCK221H00G BWTEDOCK241J00G BWTEDOCK201E00G BWTEDOCK221C1K1P00 BWTEDOCK201G00G BWTEDOCK2500G BWTEDOCK221C1M00G BWTEDOCK221C1K1N00 BWTEDOCK221J1P03G BWTEDOCK231C1K1N00 BWTEDOCK231C1K1M03 BWTEDOCK231J03G
Automatic test and calibration stations, MicroDock II
The MicroDock II are designed to bump-test, calibrate, charge and manage records for portable gas detectors. One base station with one AC outlet supports up to six modules to suit your application.
  • Verifies proper performance of detectors
  • Automatic bump testing, calibration, record keeping and charging
  • Lightweight and fully portable with optional wall mount
  • Fully customisable
  • No computer required
  • Operates via 6 V line power or a set of 4 C-cell batteries
Hard case kits: Holds 1 base station, 1 docking module and a 34 L cylinder of calibration gas, with 1 demand flow regulator, no gas.
Heavy duty case kits: Holds 1 base station, 1-3 docking module(s) and 1-3 58 L cylinder(s) of calibration gas in a foam-lined, waterproof heavy-duty case, with 1 demand flow regulator, no gas.
*Not compatible with ClO₂ or O₃.
Ordering information: Includes Fleet Manager II software for easy data analysis. Multi-language support in English, French, German, Spanish and Portuguese.