Safety helmets, Style 600
Supplier: Scott Safety
Style 600
111-2236EA 16.15 EUR
111-2236 SCTS2011259 SCTS2011287
Safety helmets, Style 600
Head Protection Safety Helmets
Style 600 offers a perfect blend of comfort, stability and balance to ensure a high degree of wearer acceptance even in the most arduous head protection environments. The helmet is made from a premium grade UV, stabilised durable ABS polymer for long lasting protection. Different headgear versions are available; optional HXSPEC EN 166 'Grade B' (120 m/s) integrated eye shield with tabs for easy retraction and deployment.
  • Side impact strength protects against lateral deformation (LD)
  • 30 mm Euro accessory slot with flat panels for easy attachment
  • Sculptured cut outs allow easy use of ear defenders
  • Terylene headgear with terry towelling sweatband
  • Rain gutter channels water off the temple
Weight: 367 to 408 g, depending on model
Certifications: EN 397