The paint inspector's field guide
SIMELD3080EA 9400 EUR
SIMELD3080
The paint inspector's field guide
The inspection of protective coating systems for corrosion control includes a wide range of test methods and techniques. The 'Paint Inspector's Field Guide', by Lee Wilson, a highly qualified and well respected inspector with many years of field experience, covers a wide range of these inspection methods in an effort to aid individuals with the basic fundamentals of protective coating inspection whilst in the field.
  • This field guide which provides a description of all the actions performed by an inspector and the tools they use, covers all aspects from specification review and surface preparation works all the way through to application and final reporting.
It is complemented by a wide range of inspection and experience notes making it easy to solve those special problems which are commonly encountered in the field. Besides the textual support, the rich graphics provide clear visual reference to inspection techniques, standards and defects.
Editing is by Brian Goldie, which makes it a very useful reference book for any individual involved or interested in corrosion control by protective coatings and the inspections required to achieve this.
Wire binded, 198 pages.