High capacity balances, Cubis® Essential
Supplier: Sartorius
SARBMSE36201SCED15EA 5400 EUR
SARBMSE36201SCED15 SARBMSE36201S-ED15 SARBMSE20201SCED15 SARBMSE36200SCED15 SARBMSE20201S-ED15
High capacity balances, Cubis® Essential
Easy operation with monolithic weighing technology for users who want to perform precise weighing.
  • MSE display - high contrast, black and white, backlit display
  • Easy to understand menu guidance wth short text prompts
  • Software support for fast levelling
  • Rectangular weighing pans
  • Three optional ports are available as accesory ( Bluetooth®, PS/2, RS232C)
Applications include unit conversion, isoCAL automatic calibration | adjustment function, density determination (buoyancy method only), calculations, averaging, net | total formulation, weighing in percent, counting, totalising.