You Searched For: Sealing Film Applicators
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (732-3335)
Supplier: VWR Collection
Description: Compact and portable, the cap mat applicator is very easy to use, requiring only one operation of the system to produce an accurate and tight seal on ...
UOM: 1 * 1 items
Catalog Number: (391-0241)
Supplier: VWR Collection
Description: Sealing film applicator, VWR®, Film sealing paddles, non sterile
UOM: 1 * 5 items
Catalog Number: (391-1278)
Supplier: VWR Collection
Description: Soft rubber roller recommended for assuring tight and consistent application of adhesive films to microplates.
UOM: 1 * 1 items
Supplier: Industrial Physics
Description: Adjustable Micrometer Film Applicator. The applicator blade can be adjusted in 10 μm increments by means of 2 μm heads, over a gap size from 0 to 8 mm...
Catalog Number: (732-0907)
Supplier: Corning
Description: Axygen® EASY PRESS is a manual device which provides simple and effective application of Axymat™ and/or Impermamat™ sealing mats to 24, 48, 96 or 384 ...
UOM: 1 * 1 items
Supplier: Industrial Physics
Description: Temperature gradient plate for determining minimum film forming temperature (MFFT) test. The instrument can be used to define the white point/glass tr...
Catalog Number: (732-1380)
Supplier: Corning
Description: Axygen® sealing film roller is a durable and efficient roller for applying sealing films to any plate. Sturdy plastic handle and semi-hard roller head...
UOM: 1 * 1 items
Catalog Number: (731-0461)
Supplier: iST Scientific Ltd
Description: This adhesive seal roller ensures even pressure is applied across the seal for a complete application to the plate.
UOM: 1 * 1 items
Catalog Number: (732-2705)
Supplier: Thermo Fisher Scientific
Description: Convenient, handheld applicator for sealing tapes onto plates.
UOM: 1 * 2 items
Catalog Number: (293-1411)
Supplier: Simport Scientific
Description: Sealing film applicator, Roller, rubber, Amplateâ„¢
UOM: 1 * 1 items
Catalog Number: (479-1248)
Supplier: MATRIX TECHNOLOGIES CORP. LTD
Description: Thermo Scientificâ„¢ SuperSealer ensures a proper seal on plates, blocks and storage tube racks.
UOM: 1 * 1 items
Catalog Number: (DIVEBRAY-4000)
Supplier: DIVERSIFIED BIOTECH
Description: Roller used to seal microplates uniformly.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom