Uracil-DNA-Glycosylase, peqGOLD
PEQLAB peqGOLD
PEQL04-1090EA 59.8 EUR
PEQL04-1090
Uracil-DNA-Glycosylase, peqGOLD
peqGOLD Uracil DNA-Glycosylase (UDG) in combination with dUTP prevents contamination in PCR reactions.
After incorporation of uracil into the DNA strand, UDG catalyses the hydrolysis of the n-glycosidic bond between uracil and deoxyribose. Before starting the PCR an incubation at 37 °C for 10 minutes degrades uracil containing DNA fragments from previous experiments. The enzyme is deactivated by heat shock at 95 °C for 10 minutes.