DNA ladder, O'range 100 bp, peqGOLD
PEQL25-2310EA 137 EUR
PEQL25-2310
DNA ladder, O'range 100 bp, peqGOLD
DNA ladder for accurate qualitative analysis of small dsDNA.
  • Bands made by ligation of defined DNA fragments
  • Contains Orange G dye instead of Bromophenol Blue
  • Range: 15 fragments, 100 – 1500 bp
  • Three highlighted bands for easy orientation
  • Ready to use mix for fast and convenient workflow