DNA ladder, O'range 50 bp, peqGOLD
PEQL25-2300EA 124 EUR
PEQL25-2300
DNA ladder, O'range 50 bp, peqGOLD
DNA ladder for accurate qualitative analysis of small dsDNA.
  • Bands made by ligation of defined DNA fragments
  • Contains Orange G dye instead of Bromophenol Blue
  • Range: 20 fragments, 50 – 1000 bp
  • Three highlighted bands for easy orientation
  • Ready to use mix for fast and convenient workflow