DNA ladder, low range, peqGOLD
PEQL25-2220EA 156 EUR
PEQL25-2220
DNA ladder, low range, peqGOLD
Low range DNA ladder for accurate quantitative and qualitative analysis of small dsDNA.
  • Quantity of each fragment is accurately calibrated
  • Range: 11 fragments, 80 to 1031 bp
  • Highlighted 500 bp band for easy orientation
  • Ready to use for fast and convenient workflow