DNA Sizer XII, peqGOLD
PEQL25-2160EA 0 EUR
PEQL25-2160
DNA Sizer XII, peqGOLD
Classic pUC19 DNA sizer for quantitative and qualitative analysis of small dsDNA.
  • pUC19 plasmid was digested with Hpa II and purified
  • Range: 13 fragments, 26 to 501 bp
  • 6X loading buffer included