XChange Plasmid Maxi Kit, peqGOLD
PEQLEMR509010EA 110 EUR
PEQLEMR509010 PEQLEMR509020 PEQL12-7402-02
XChange Plasmid Maxi Kit, peqGOLD
XChange Plasmid Maxi Kit for the isolation of up to 520 µg plasmid DNA per preparation. Anion exchange technology ensures highly pure and intact plasmids up to 300 kbp. Plasmids isolated with the peqGOLD XChange Plasmid Maxi kit are highly pure and can directly be used for a wide range of downstream applications.
  • Quick and easy protocol - short hands-on and preparation times of only 80 minutes
  • No need for centrifugation steps effectively protects DNA during extraction
  • Complete enzymatic degradation of all cellular RNA and nucleases
  • Efficient removal of enzyme inhibitors and contaminants
  • Plasmid DNA is suitable for high transfection efficiencies
Delivery information: Each kit includes XChange Maxi columns, folded filters, plastic washers, Solution I, II and III, EQ Buffer, DNA Wash Buffer, RNase A, and Elution Buffer.