Bacterial RNA Kit, peqGOLD
PEQL12-6850-00EA 27.1 EUR
PEQL12-6850-00 PEQL12-6850-01 PEQL12-6850-02
Bacterial RNA Kit, peqGOLD
Bacterial RNA Kit for the extraction of high quality total RNA from gram+ or gram– bacteria. RNA extractions using this kit are quick, cheap and achieve reliable results.
  • Also suited for extractions from difficult species
  • Suitable for the simultaneous processing of multiple samples
  • Immediate inactivation of endogenous and exogenous RNases
  • No need for organic extractions
  • Efficient separation of enzyme inhibitors and contaminants
  • Isolated RNA is directly suitable for downstream applications by elution in RNase-free water
* Safety Line columns with snap-on lids.
Delivery information: Each kit contains PerfectBind RNA columns (Safety Line*), collection tubes, DNA Removing columns, RNA Lysis Buffer T, RNA Wash Buffer I and II, and RNase-free water.