You Searched For: Sample Cups for Analysers
16  results were found
SearchResultCount:"16"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: KIMA SAS DI CHIARIN RENZO & C
Description: Cups made of PS for auto analyser.
Supplier: KARTELL
Description: PS, transparent.
Supplier: Brand
Description: PS, transparent.
Supplier: Industrial Physics
Description: Permeability cups are designed to determine the water vapour transmission of paints, varnishes, coatings and related products. Permeability cup is sui...
Catalog Number: (SIMPB720-8)
Supplier: Simport Scientific
Description: These analyser cups are made of PS.
UOM: 1 * 10.000 items
Supplier: Simport Scientific
Description: PS, clear.
Supplier: PerkinElmer
Description: Disposable.
Supplier: RATIOLAB
Description: Made of clear PS.
Catalog Number: (720-0812)
Supplier: KARTELL
Description: Polystyrene cup with polyethylene lid graduated at 10 and 20 ml.
UOM: 1 * 200 items
Catalog Number: (METB00018127)
Supplier: Mettler - Toledo
Description: Kit, Set of 7 PC nipples
UOM: 1 * 7 items
Catalog Number: (DELT200103)
Supplier: DELTALAB
Description: Single use PS cups suitable for cell counting.
UOM: 1 * 1.000 items
Supplier: DELTALAB
Description: Cups made of PS.
Catalog Number: (SIMPB720-40)
Supplier: Simport Scientific
Description: PE, translucent.
UOM: 1 * 1.000 items
Catalog Number: (391-3720)
Supplier: Greiner Bio-One
Description: Cups made of PS.
UOM: 1 * 1.250 items
Supplier: SARSTEDT
Description: Sample cups, made of PS.
Catalog Number: (SARS73.663)
Supplier: SARSTEDT
Description: PS, transparent.
UOM: 1 * 5.000 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 16
no targeter for Bottom