Dust and fume extraction system, Cleantec® LEV
Supplier: Bigneat
BIGNCT0200/01EA 1680 EUR
BIGNCT0200/01
Dust and fume extraction system, Cleantec® LEV
Cleantec LEV system is designed to solve fume, dust and odour problems. Efficient filtration within a powerful extraction system removes hazardous material from process direct from where the extraction is required. This system is adaptable and may be used as a fully mobile system on a mobile workbench, with a handle and on castors and with a single flexible or jointed arm, or maybe installed under bench with multiple arms or connected direct into process equipment.
  • Easily changed pre-filters give extended HEPA filter life
  • Portable and adaptable to your changing process requirements
  • Standard electrical socket connection for fan filter module supply
Ordering information: Accessories, nozzles, cowls and arms are available on request.