VWR®, Ready PCR Mix, 2X
Supplier: Avantor
VWR®
N806-2X1.25MLEA 122 EUR
N806-2X1.25ML
VWR®, Ready PCR Mix, 2X
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Ready PCR Mix, 2X offers a single-step procedure for performing PCR reactions followed by analysis on agarose gels without the addition of loading buffer. All components for assembly and performance of PCR reactions as well as loading and visualisation of PCR products on agarose gels are included. The user supplies primer and template DNA.
  • Direct to gel loading
  • Immediate visualisation
  • Safe to use