Anti-H2B Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
CAYM13538-1EA 458 EUR
CAYM13538-1
Anti-H2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: H2B
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Dog, Drosphila, Human, most mammals, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR