Anti-Insulin Mouse Monoclonal Antibody [clone: 2D11-H5]
Supplier: United States Biological
USBI138556-100EA 608 EUR
USBI138556-100 USBI138556-20
Anti-Insulin Mouse Monoclonal Antibody [clone: 2D11-H5]
Antibodies
Recommended dilution: ;Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: Insulin
Clonality: Monoclonal
Clone: 2D11-H5
Conjugation: unconjugated
Epitope: full-length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Human, Pig,