Anti-LAT2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA322392EA 564 EUR
ORIGTA322392
Anti-LAT2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:10-50

Type: Primary
Antigen: LAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR