You Searched For: Atomisers
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (217-0695)
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass sprayer.
UOM: 1 * 1 items
Catalog Number: (MFLX70025-04)
Supplier: Avantor Fluid Handling
Description: Increase oxygen into your water.
UOM: 1 * 1 items
Supplier: PerkinElmer
Description: The PFA-ST MicroFlow nebuliser is made from high purity, HF resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake ...
Catalog Number: (CAMA022.6530)
Supplier: CAMAG
Description: Reagents are atomised into a fine aerosol spray with particles in the range of 0,3 to 10 µm.
UOM: 1 * 1 items
Catalog Number: (552-0031)
Supplier: CAMAG
Description: Glass atomiser. Cost efficient alternative for a TLC sprayer.
UOM: 1 * 1 items
Catalog Number: (552-6001)
Supplier: WITEG LABORTECHNIK GMBH
Description: Glass spray bottle with rubber bulb for simple manual operation of the solution spray.
UOM: 1 * 1 items
Catalog Number: (HEWLG1820-65337)
Supplier: Agilent
Description: This spray chamber made of quartz is used with 7500 ICP-MS.
UOM: 1 * 1 items
Catalog Number: (HEWLG3139-65003)
Supplier: Agilent
Description: This PFA spray chamber is the standard spray chamber used with the 7500 ICP-MS when aspirating hydrofluoric acid.
UOM: 1 * 1 items
Catalog Number: (WITG2.147.000)
Supplier: WITEG LABORTECHNIK GMBH
Description: Sprayer are made of borosilicate glass, complete with rubber bulb.
UOM: 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Description: These test tube sprayers are made of DURAN®, borosilicate glass.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom