You Searched For: Injection Valves
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SGE a Trajan company
Description: Syringe valve VLLMA, push button with Luer lock, 5 ml - 2 l syringe volume
Supplier: IDEX HEALTH&SCIENCE LLC(RHEODY
Description: Manual sample injectors have two positions, 'Load' and 'Inject'. The 'Load' position allows sample loading into the sample loop or sample chamber. A s...
Catalog Number: (HAMI86580)
Supplier: HAMILTON BONADUZ
Description: This MTB sampling valve is a lever-actuated on/off valve used for storing and transporting samples in any Gastight® PTFE Luer lock (TLL) or Luer tip (...
UOM: 1 * 1 items
Catalog Number: (WHEA851426)
Supplier: DWK Life Sciences
Description: Kit includes 1 spring, 2 balls, 2 washers for valve, and barrel washer.
UOM: 1 * 1 items
Supplier: SOCOREX ISBA
Description: Replacement kits for the valve assembly are supplied in a dedicated bag with each Socorex® self-refilling syringe.
Supplier: Agilent
Description: Replacement valve WE series, 10-port, 400 psi, 225 °C
Catalog Number: (WITG0.290.403)
Supplier: WITEG LABORTECHNIK GMBH
Description: Spare needle valve, PTFE, red for high vacuum stopcocks WITAFLO, 3 mm hole
UOM: 1 * 1 items
Supplier: Agilent
Description: The stainless steel 7725i and PEEK 9725i manual injection valves are suitable for large sample volumes, high flow rates and preparative columns sized ...
Supplier: Restek
Description: Sulfinert® treated gas sample loops are ideal for samples containing low concentrations of sulphur and other active compounds.
Supplier: Merck
Description: The sampling valves are used for sterile sampling of water or other final products in the pharmaceutical industry.
Catalog Number: (560-0168)
Supplier: UPCHURCH SCIENTIFIC
Description: The 6-port medium pressure injection valves feature easy, external sample loop customisation. Simply clean-cut a piece of tubing that represents the v...
UOM: 1 * 1 items
Catalog Number: (SIAL57620-U)
Supplier: Merck
Description: The Rheodyne Model 7725 injector allows you to inject 1 μl to 5 ml samples with accuracy and precision.
UOM: 1 * 1 Each
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom