Antibiotic assay strips
Supplier: Oxoid
OXOIMA0113FEA 0 EUR
OXOIMA0113F OXOIMA0101F OXOIMA0100F OXOIMA0122F OXOIMA0121F OXOIMA0105F OXOIMA0104F OXOIMA0118F
Antibiotic assay strips
The M.I.C.Evaluator™ (M.I.C.E.™) strips are a range of devices for the accurate determination of the minimum inhibitory concentration (MIC) of a test organism to an antimicrobial. The M.I.C.E. strips consist of a gradient of stabilised antimicrobial covering 15 doubling dilutions. The innovative new design makes the clinical interpretation of the MIC even easier by removing the half step values. The accuracy of the test is still maintained if required using the black sections on the strip. On application of a M.I.C.E. strip to a pre-inoculated agar plate, the antimicrobial releases from the polymer strip, forming a defined concentration gradient in the area around it. After appropriate incubation, growth develops with a zone of inhibition around the strip.
  • Easy to read
  • Individually sealed in an easily peelable sachet with a desiccant to maintain the long term stability of the product