Cooling carrier, BioT™
BCISBCS-528EA 1150 EUR
BCISBCS-528
Cooling carrier, BioT™
BioT™ Carrier is a portable 2 - 8 °C cooling solution for transporting small batches of temperature-sensitive laboratory research samples. It maintains and transports the cold samples with confidence. Easily adjust the cooling temperature set point to raise or lower the chamber temperature to best suit your application. It carries cold samples between facilities, from clinic to lab or over longer distances utilizing the car power adaptor.
  • Intuitive LCD display
  • Rechargeable battery lasts up to 8 hours
  • Audible alarm when battery is low
  • Magnetized lid
  • Rugged and lightweight
Weight: 5,3 kg
Delivery information: BioT™ Carrier includes AC power adaptors with plug, rechargeable battery, car power adaptor, shoulder strap and accessories bag.