Live cell analysers, JuLI™ FL
Supplier: NanoEnTek
JuLI™ Fl
NANOJULI-FGSCEA 11010 EUR
NANOJULI-FGSC 734-3200 734-3201 734-3202 734-3203 NANOJULI-FRSC
Live cell analysers, JuLI™ FL
Cell Counters and Flow Cytometers Automated Cell Counters
The JuLI Fl live cell analysers offers all the features as the JuLI Br brightfield but with the additional capability to analyse fluorescently labelled cells: The microscope is placed inside the cell culture incubator, the base station remains on the laboratory bench.
  • Data formats: JPEG, TIFF, BMP, PNG (images), AVI (video), CSV (raw data)
  • Connection of an additional microscope unit to the base station for the direct comparison of two samples
  • Microscope unit and light-sensitive digital camera that utilises either a GFP or RFP channel for fluorescence detection
Applications include cell growth monitoring, creation of time-lapse images and movies, cell confluence detection, recording cell growth in real-time, wound healing (scratch) assays, and cell counting.