Reduced-serum medium, transfectagro™
Supplier: Corning Life Sciences
Mediatech
40-300-CVEA 177.1 EUR
40-300-CV
Reduced-serum medium, transfectagro™
Cell Culture Media
transfectagro™ Reduced-serum medium is designed to maximise the growth of a variety of cell types under reduced-serum conditions, generally allowing for at least a 50% reduction in serum use with minimal adaptation. A chemically defined, reduced-serum formulation, transfectagro™ also improves transient transfection efficiency in a variety of cell types.