Anti-STK35 Goat Polyclonal Antibody

Supplier: United States Biological
USBIS7973-58Z4EA 551 EUR
USBIS7973-58Z4
Anti-STK35 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: STK35 (serine/threonine kinase 35)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR