Anti-PRMT3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Supplier: United States Biological
USBIP9004-17B-APEA 852 EUR
USBIP9004-17B-AP
Anti-PRMT3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: PRMT3 (protein arginine methyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human