Anti-P2X3 Receptor Guinea pig Polyclonal Antibody

Supplier: United States Biological
USBIP1003-08HEA 627 EUR
USBIP1003-08H
Anti-P2X3 Receptor Guinea pig Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 1:500;

Type: Primary
Antigen: P2X3 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR