Anti-CD28 Mouse Monoclonal Antibody [clone: ANC28.1/5D10]

Supplier: United States Biological
USBIC2380-01LEA 520 EUR
USBIC2380-01L
Anti-CD28 Mouse Monoclonal Antibody [clone: ANC28.1/5D10]
Antibodies
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: Monoclonal
Clone: ANC28.1/5D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR