Anti-CD3E Mouse Monoclonal Antibody [clone: 5H55]

Supplier: United States Biological
USBIC2254-61EA 571 EUR
USBIC2254-61
Anti-CD3E Mouse Monoclonal Antibody [clone: 5H55]
Antibodies
Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: Monoclonal
Clone: 5H55
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR