Anti-CBLB Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Supplier: United States Biological
USBIC0013-32B-APCEA 616 EUR
USBIC0013-32B-APC
Anti-CBLB Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: CBLB (Cbl proto-oncogene B, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human