Anti-BGN Sheep polyclonal antibody unconjugated

Supplier: United States Biological
USBIB1089-40BEA 641 EUR
USBIB1089-40B
Anti-BGN Sheep polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: BGN (biglycan)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR