Anti-AKT2 Mouse Monoclonal Antibody [clone: 1B6]

Supplier: United States Biological
USBIA1125-26FEA 633 EUR
USBIA1125-26F
Anti-AKT2 Mouse Monoclonal Antibody [clone: 1B6]
Antibodies
Recommended dilution: Immunofluorescence: 1:200-1:1000; ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:100

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: monoclonal
Clone: 1B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR