Anti-AKT2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: United States Biological
USBIA1125-25A-APEA 852 EUR
USBIA1125-25A-AP
Anti-AKT2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR